DWK73800-15415
KIMBLE® Solid Cap
with rubber liner, phenolic, cap for 15-415
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
PTFE/rubber liner
sterility
non-sterile
feature
class A
packaging
case of 1000 ea
manufacturer/tradename
Kimble® 73800-15415
fitting
cap for 15-415
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavable
- White rubber liners
- Excellent for general laboratory use
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)