DWK223747
WHEATON® Type I crimp top serum bottle
without cap, round clear borosilicate glass bottle, capacity (100 mL)
Synonym(s):
223747
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
round clear borosilicate glass bottle
feature
without cap
packaging
case of 144 ea Shrink-wrapped partitioned modules
manufacturer/tradename
WHEATON® 223747
bottle diam. × H × W
(52 mm) (95 mm) (337 mm)
capacity
(100 mL)
Looking for similar products? Visit Product Comparison Guide
General description
Serum Bottles with exceptional wall strength and superior heat and chemical tolerances.
- Ideal for long and short term sample storage, lyophilization and vaccine / injectable drug containers
- Fits most lyophilization applications
- Manufactured from WHEATON 400 borosilicate molded glass that conforms to USP Type I requirements
- Shrink-wrapped partitioned modules reduce breakage
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service