Key Documents
DWK14395-100
KIMBLE® KIMAX® Media bottle
with cap, round transparent borosilicate glass bottle, bottle capacity (100 mL), non-sterile
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
bottle capacity:
(100 mL)
material:
blue polypropylene cap
round transparent borosilicate glass bottle
sterility:
non-sterile
feature:
with cap
fitting:
GL 45 neck
GL 45
packaging:
case of 10 ea
material
blue polypropylene cap
round transparent borosilicate glass bottle
sterility
non-sterile
feature
with cap
packaging
case of 10 ea
manufacturer/tradename
Kimble® 14395-100
bottle capacity
(100 mL)
fitting
GL 45 neck
GL 45
General description
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Features and Benefits
• Enhanced graduations and marking spot made with chemically resistant white enamel paint• 30 mm ID opening• Autoclavable• Supplied without caps or with linerless GL 45 screw thread caps• Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)