Skip to Content
Merck
All Photos(3)

Key Documents

D218200

Sigma-Aldrich

5,5′-Dithiobis(2-nitrobenzoic acid)

ReagentPlus®, 99%

Synonym(s):

3-Carboxy-4-nitrophenyl disulfide, 6,6′-Dinitro-3,3′-dithiodibenzoic acid, Bis(3-carboxy-4-nitrophenyl) disulfide, DTNB, Ellman’s Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SC6H3(NO2)CO2H]2
CAS Number:
Molecular Weight:
396.35
Beilstein:
1896821
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

mp

240-245 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(SSc2ccc(c(c2)C(O)=O)[N+]([O-])=O)ccc1[N+]([O-])=O

InChI

1S/C14H8N2O8S2/c17-13(18)9-5-7(1-3-11(9)15(21)22)25-26-8-2-4-12(16(23)24)10(6-8)14(19)20/h1-6H,(H,17,18)(H,19,20)

InChI key

KIUMMUBSPKGMOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DTNB (Ellman’s reagent) is commonly used for quantification of sulfhydryl (thiol) groups in proteins and biomolecules.

Application

5,5′-Dithiobis(2-nitrobenzoic acid) is a water soluble reagent mainly used for the derivatization of sulfhydryl groups.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pork proteins oxidative modifications under the influence of varied time-temperature thermal treatments: A chemical and redox proteomics assessment.
Mitra B, et al.
Meat Science, 140, 134-144 (2018)
Determination of thiols and disulfides via HPLC quantification of 5-thio-2-nitrobenzoic acid
W Chen, et al.
Journal of Pharmaceutical and Biomedical Analysis, 48, 1375-1380 (2008)
Atef M K Nassar et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-11)
Sumithion (Fenitrothion) (SUM) is an organophosphorus insecticide used to combat a wide variety of plant pests. Exposure to SUM causes significant toxicity to the brain, liver, kidney, and reproductive organs through, for example, binding to DNA, and it induces DNA
Anne Bertling et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(8), 1979-1990 (2012-04-28)
Staphylococcus aureus can induce platelet aggregation. The rapidity and degree of this correlates with the severity of disseminated intravascular coagulation, and depends on platelet peptidoglycans. Surface-located thiol isomerases play an important role in platelet activation. The staphylococcal extracellular adherence protein
Malgorzata Boczkowska et al.
Structure (London, England : 1993), 16(5), 695-704 (2008-05-09)
Previous structures of Arp2/3 complex, determined in the absence of a nucleation-promoting factor and actin, reveal its inactive conformation. The study of the activated structure has been hampered by uncontrollable polymerization. We have engineered a stable activated complex consisting of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service