D182001
3,5-Dimethylpyrazole
99%
Synonym(s):
3,5-Dimethyl-1H-pyrazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
Beilstein:
106325
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39160503
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
bp
218 °C (lit.)
mp
105-108 °C (lit.)
SMILES string
Cc1cc(C)[nH]n1
InChI
1S/C5H8N2/c1-4-3-5(2)7-6-4/h3H,1-2H3,(H,6,7)
InChI key
SDXAWLJRERMRKF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Common reagent for the preparation of pyrazolato ligated complexes. Also used to prepare N-1-substituted derivatives having antibacterial activity.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2
Target Organs
Liver
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of Organometallic Chemistry, 443, 221-221 (1993)
J. Chem. Soc., Dalton Trans., 3625-3625 (1993)
J. Chem. Soc. Pak., 14, 125-125 (1992)
E Bergamini et al.
The American journal of physiology, 266(1 Pt 1), G118-G122 (1994-01-01)
Regulation of liver macroautophagy and protein degradation by hormones and direct regulatory amino acids were studied in male 2-mo-old Sprague-Dawley albino rats with the use of the antilipolytic agent 3,5'-dimethylpyrazole (DMP; 12 mg/kg body wt ip) as a stimulatory agent.
C Redekopp et al.
Journal of endocrinological investigation, 7(4), 277-281 (1984-08-01)
The inhibitory and stimulatory effects of somatostatin analogues on growth hormone secretion have been studied in the sheep. Plasma immunoreactive growth hormone (GH) was stimulated by the iv administration of the antilipolytic compound 3,5-dimethylpyrazole and was followed by infusion of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service