D153303
5,5-Dimethyl-1,3-cyclohexanedione
95%
Synonym(s):
Dimedone, Methone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
95%
form
powder
mp
146-148 °C (lit.)
SMILES string
CC1(C)CC(=O)CC(=O)C1
InChI
1S/C8H12O2/c1-8(2)4-6(9)3-7(10)5-8/h3-5H2,1-2H3
InChI key
BADXJIPKFRBFOT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Part of furannulation strategy for the synthesis of naturally occurring fused 3-methylfurans.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of Organic Chemistry, 58, 3960-3960 (1993)
Methods in enzymology, 473, 117-136 (2010-06-02)
Protein sulfenic acids (R-SOH) are receiving increased interest as intermediates in redox processes. Human serum albumin, the most abundant protein in plasma, possesses a single free thiol. We describe herein the different methodologies that we have employed to study the
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 83(6), 110-115 (2012-03-01)
A method for endogenous formaldehyde (FA) level evaluation has been worked out. The method involves the administration of dimedone, which forms the stable complex with FA, and the determination of formaldimedone concentration in biological samples by the fluorescence approach. The
Journal of natural products, 74(4), 757-763 (2011-02-24)
Sphaeropsidone and episphaeropsidone are two phytotoxic dimedone methyl ethers produced by Diplodia cupressi, the causal agent of a canker disease of cypress in the Mediterranean area. In this study, eight derivatives obtained by chemical modifications and two natural analogues were
BMC biochemistry, 11, 25-25 (2010-07-03)
Cysteine sulfenic acid (Cys-SOH) plays important roles in the redox regulation of numerous proteins. As a relatively unstable posttranslational protein modification it is difficult to quantify the degree to which any particular protein is modified by Cys-SOH within a complex
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service