D122807
1,2-Diiodoethane
99%
Synonym(s):
Ethylene diiodide, Ethylene iodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ICH2CH2I
CAS Number:
Molecular Weight:
281.86
Beilstein:
1730870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder or crystals
mp
80-82 °C (lit.)
density
2.132 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
ICCI
InChI
1S/C2H4I2/c3-1-2-4/h1-2H2
InChI key
GBBZLMLLFVFKJM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1,2-Diiodoethane can be used:
- To synthesize samarium(II) iodide-water complex (SmI2-H2O complex), a single-electron transfer reagent, to further synthesize 3-hydroxy carboxylic acids.
- To synthesize derivatives of regioselective homopropargyl alcohols by using sonochemical Barbier-type reaction conditions.
- As an iodine source and an effective reagent for the dehydroxy-iodination of alcohols.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of homopropargyl alcohols via sonochemical Barbier-type reaction.
Lee ASY, et al.
Tetrahedron Letters, 45(7), 1551-1553 (2004)
Halogenation through Deoxygenation of Alcohols and Aldehydes.
Chen J, et al.
Organic Letters, 20(10), 3061-3064 (2018)
Michal Szostak et al.
Nature protocols, 7(5), 970-977 (2012-04-28)
The single-step synthesis of 3-hydroxy carboxylic acids from readily available Meldrum's acids involves a selective monoreduction using a SmI(2)-H(2)O complex to give products in high crude purity, and it represents a considerable advancement over other methods for the synthesis of
Jiajun Yan et al.
Nature nanotechnology, 14(7), 684-690 (2019-05-22)
Eutectic gallium indium (EGaIn) is a liquid metal alloy at room temperature. EGaIn microdroplets can be incorporated into elastomers to fabricate highly stretchable, mechanically robust, soft multifunctional composites with high thermal stability1 and electrical conductivity2-4 that are suitable for applications
Eugen Klein et al.
ACS nano, 13(6), 6955-6962 (2019-06-12)
Hybrid lead halide perovskites with 2D stacking structures have recently emerged as promising materials for optoelectronic applications. We report a method for growing 2D nanosheets of hybrid lead halide perovskites (I, Br and Cl), with tunable lateral sizes ranging from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service