CLS3026100
Pyrex® class A double metric scale graduated cylinder, with blue enamel
volume 100 mL
Synonym(s):
Corning® 3026
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
subdivision 1.0 mL
manufacturer/tradename
Corning 3026-100
tol. measuring range
0.50 mL
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Calibrated "to deliver".
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service