CLS15001L
Corning® narrow mouth reagent bottle with Pyrex® ST stopper
capacity 1000 mL, stopper joint: 29
Synonym(s):
glass bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
clear bottle
cylindrical bottle
narrow-mouth bottle
Joint
joint: stopper29
feature
cap
manufacturer/tradename
Corning 1500-1L
capacity
1000 mL
diam. × H
107 mm × 200.5 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
These Pyrex narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service