Key Documents
CLS139545DC
Delivery cap for Corning® media storage bottles
red
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
manufacturer/tradename
Corning 1395-45DC
color
red
General description
This three hole delivery cap has 3 holes with 1/4-28 threads at the top for use with 1/8in. O.D. tubing. No need to precut tubing to reach the reservoir bottom, just a quick quarter turn of the lock nut (from outside the reservoir) allows positioning of PTFE tubing at any depth. These delivery caps have a GL 45 thread, can be dry heat sterilized at up to 180 °C and have a PTFE liner for efficient sealing.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)