Skip to Content
Merck
All Photos(1)

Key Documents

CDS021533

Sigma-Aldrich

Cyclopentolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25NO3
CAS Number:
Molecular Weight:
291.39
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

OC1(CCCC1)C(C(OCCN(C)C)=O)C2=CC=CC=C2

InChI

1S/C17H25NO3/c1-18(2)12-13-21-16(19)15(14-8-4-3-5-9-14)17(20)10-6-7-11-17/h3-5,8-9,15,20H,6-7,10-13H2,1-2H3

InChI key

SKYSRIRYMSLOIN-UHFFFAOYSA-N

Gene Information

human ... CHRM3(1131)

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Tian Kang et al.
Clinical & experimental ophthalmology, 44(8), 701-709 (2016-04-16)
To report the thickness of the peripapillary retinal nerve fibre layer (pRNFL) in Chinese children and examine its association with refractive error, axial length (AL) and optic disc parameters. Population-based cross-sectional study. A total of 2893 seven-year-old children from 11
Glenn Yiu et al.
Scientific reports, 7(1), 15013-15013 (2017-11-05)
Nonhuman primates are the only mammals to possess a true macula similar to humans, and spontaneously develop drusenoid lesions which are hallmarks of age-related macular degeneration (AMD). Prior studies demonstrated similarities between human and nonhuman primate drusen based on clinical
Xiao Nicole Liu et al.
Scientific reports, 10(1), 17194-17194 (2020-10-16)
Myopia, a leading cause of distance vision impairment, is projected to affect half of the world's population in 30 years. We analysed the relationship between certain demographic, environmental, and behavioural factors and myopia from a 2-year school-based, prospective trial conducted in
Yan Li et al.
BMC ophthalmology, 17(1), 88-88 (2017-06-14)
Myopia is a leading cause of preventable blindness. Although, multiple cross-sectional epidemiological studies have confirmed that there is a high prevalence of myopia in high school-aged students in China. However, few longitudinal studies have been performed to assess junior high
Chen-Wei Pan et al.
Clinical & experimental optometry, 101(6), 758-763 (2018-03-27)
To determine the associations of reduced presenting visual acuity (VA), refractive errors and health-related quality of life (HRQoL) in healthy adolescents in rural China. A total of 2,235 grade 7 students without systemic medical problems from 10 middle schools aged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service