Skip to Content
Merck
All Photos(1)

Documents

C56255

Sigma-Aldrich

Chloromethylphosphonic dichloride

97%

Synonym(s):

(Chloromethyl)phosphonic acid dichloride, (Chloromethyl)phosphonic dichloride, (Chloromethyl)phosphonoyl dichloride, (Chloromethyl)phosphoryl dichloride, Chloromethanephosphonic acid dichloride, Chloromethanephosphonic dichloride, Chloromethyl dichlorophosphine oxide, Chloromethylphosphonyl dichloride, Dichloro(chloromethyl)phosphine oxide, P-(Chloromethyl)phosphonic dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2P(O)Cl2
CAS Number:
Molecular Weight:
167.36
Beilstein:
969707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

78-82 °C/10 mmHg (lit.)

density

1.638 g/mL at 25 °C (lit.)

SMILES string

ClCP(Cl)(Cl)=O

InChI

1S/CH2Cl3OP/c2-1-6(3,4)5/h1H2

InChI key

FZXSSJIKXCFPDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service