C116807
Cyclopropanecarbonyl chloride
98%
Synonym(s):
Cyclopropanecarboxylic acid chloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C3H5COCl
CAS Number:
Molecular Weight:
104.53
Beilstein:
471286
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.452 (lit.)
bp
119 °C (lit.)
density
1.152 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
ClC(=O)C1CC1
InChI
1S/C4H5ClO/c5-4(6)3-1-2-3/h3H,1-2H2
InChI key
ZOOSILUVXHVRJE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Cyclopropanecarbonyl chloride can be used as an alkylating reagent in the preparation of some chemical building blocks such as formylcyclopropane, cyclopropanecarboxylic acid hydrazide, bis(cyclopropylcarbonyl) peroxide, 1-cyclopropanecarboxamide, 2-cyclopropylcarbonylcyclohexane-1,3-diones and cyclopropyl analog of triazolopiperazine-amides.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Enzyme inhibition potency enhancement by active site metal chelating and hydrogen bonding induced conformation-restricted cyclopropanecarbonyl derivatives
Kuo P-Y, et al.
Bioorganic & Medicinal Chemistry Letters, 16(23), 6024-6027 (2006)
Palladium (II)-catalyzed highly enantioselective C-H arylation of cyclopropylmethylamines
Chan Kelvin SL, et al.
Journal of the American Chemical Society, 137(5), 2042-2046 (2015)
Preparation of Pivaloyl Hydrazide in Water: (Propanoic acid, 2, 2-dimethyl-, hydrazide)
Li Bryan, et al.
Organic Syntheses, 81(23), 254-261 (2003)
Thermal decomposition of diacyl peroxide. Part 10. Evidence for acyloxyl radical pair mechanism for 18 O-scrambling of 18 O-labelled cyclopropanecarbonyl peroxide
Fujimori K, et al.
J. Chem. Soc. Perkin Trans. II, 21(3), 405-412 (1996)
Triazolopiperazine-amides as dipeptidyl peptidase IV inhibitors: Close analogs of JANUVIA?(sitagliptin phosphate)
Kim, Dooseop, et al.
Bioorganic & Medicinal Chemistry Letters, 17(12), 3373-3377 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service