BR90438
BRAND® sampling dipper with spout
capacity 100 mL, D × H 55 mm × 65 mm
Synonym(s):
PTFE, Sampling dippers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104118
NACRES:
NB.51
Recommended Products
material
PTFE
manufacturer/tradename
BRAND 90438
D × H
55 mm × 65 mm
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
With reinforced rim and spout. PTFE handle with steel core.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service