BR759500
BRAND® cuvette rack
Holds 16 cuvettes
Synonym(s):
Accessories, Cuvettes, Racks
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
gray
polypropylene
feature
Holds 16 cuvettes
packaging
pack of 1 ea
manufacturer/tradename
BRAND 759500
size
210 mm × 70 mm × 38 mm
Related Categories
General description
Our Grey PP Cuvette Rack is designed for efficient organization and storage of cuvettes in laboratory settings. Featuring numbered positions for easy identification, this rack measures 210 x 70 x 38 mm, making it compact yet functional. Constructed from durable polypropylene (PP), it is autoclavable at 121 °C, ensuring thorough sterilization for safe use. This cuvette rack is an essential tool for maintaining order and accessibility in your laboratory, enhancing workflow and productivity.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)