Skip to Content
Merck
All Photos(1)

Key Documents

BR30616

BRAND® USP BLAUBRAND® volumetric pipettes, one-mark

capacity 30 mL, class AS, USP, DE-M

Synonym(s):

BLAUBRAND®, Bulb pipettes, USP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

AR-Glass

feature

class AS USP, DE-M
DE-M marking

packaging

pack of 6 ea

manufacturer/tradename

BRAND 30616

capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® USP BLAUBRAND® volumetric pipettes, one-mark, are crafted from high-quality AR-Glas® and comply with DIN EN ISO 648 standards. These pipettes are adjusted to "Ex" and feature a wait time of 5 seconds for precise liquid delivery. Each pipette includes a USP batch certificate, ensuring quality assurance and traceability. This reliable volumetric pipette is essential for accurate liquid measurement in various laboratory applications, making it a valuable tool for precise analytical work.

Features and Benefits

  • BRAND® USP BLAUBRAND® 30 mL one mark volumetric pipette is made of chemically resistant glass for durability and precision.
  • Computer-controlled, automated production line ensures maximum precision.
  • High contrast blue enamel print represents the optimum combination of resistance and readability.
  • The optimized flow of geometry in the tip ensures reliable liquid dispensing from the pipette.
  • The mechanical resilience of the tip and its fire-polished edges ensures a long service life.
  • Calibrated to deliver (TD, Ex).
  • Wait time of 5 seconds.

Legal Information

AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page contains detailed information on working with and handling pipettes and pipetting aids.

This page contains detailed information on working with and handling pipettes and pipetting aids.

This page contains detailed information on working with and handling pipettes and pipetting aids.

This page contains detailed information on working with and handling pipettes and pipetting aids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service