Skip to Content
Merck
All Photos(1)

Key Documents

BBO000159

Sigma-Aldrich

4-Amino-7-chloroquinoline

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClN2
CAS Number:
Molecular Weight:
178.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

Nc1ccnc2cc(Cl)ccc12

InChI

1S/C9H7ClN2/c10-6-1-2-7-8(11)3-4-12-9(7)5-6/h1-5H,(H2,11,12)

InChI key

NDRZSRWMMUGOBP-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of BioBlocks

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly Yearick et al.
Journal of medicinal chemistry, 51(7), 1995-1998 (2008-03-19)
Systematic variation of the branching and basicity of the side chain of chloroquine yielded a series of new 7-chloro-4-aminoquinoline derivatives exhibiting high in vitro activity against four different strains of P. falciparum. Many of the compounds tested showed excellent potency
Igor Opsenica et al.
Journal of medicinal chemistry, 51(19), 6216-6219 (2008-09-09)
The synthesis of the chimeric molecules consisting of two pharmacophores, tetraoxane and 7-chloro-4-aminoquinoline, is reported. The tetraoxanes 2, 4, and 8 show relatively potent in vitro antimalarial activities, with IC90 values for the Plasmodium falciparum strain W2 of 2.26, 12.44
Manolo Casagrande et al.
Bioorganic & medicinal chemistry, 18(18), 6625-6633 (2010-08-28)
A set of nine new arylpyrrolyl derivatives of 7-chloro-4-aminoquinoline, characterized by different substituents on the phenyl ring or different distance between the pyrrolic nitrogen and the 4-aminoquinoline, has been synthesized and tested for their activity against D-10 (CQ-S) and W-2
Manolo Casagrande et al.
Bioorganic & medicinal chemistry, 20(19), 5965-5979 (2012-08-25)
With the aim to investigate the effect of different heterocyclic rings linked to the 4-aminoquinoline nucleus on the antimalarial activity, a set of 7-chloro-N-(heteroaryl)-methyl-4-aminoquinoline and 7-chloro-N-(heteroaryl)-4-aminoquinoline was synthesized and tested in vitro against D-10 (CQ-S) and W-2 (CQ-R) strains of
Chiara Rusconi et al.
Bioorganic & medicinal chemistry, 20(19), 5980-5985 (2012-08-21)
Recently the N-(-)-lupinyl-derivative of 7-chloro-4-aminoquinoline ((-)-AM-1; 7-chloro-4-{N-[(1S,9aR)(octahydro-2H-quinolizin-1-yl)methyl]amino}quinoline) showed potent in vitro and in vivo activity against both Chloroquine susceptible and resistant strains of Plasmodium falciparum. However, (-)-AM-1 is synthesized starting from (-)-lupinine, an expensive alkaloid isolated from Lupinus luteus whose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service