Skip to Content
Merck
All Photos(1)

Key Documents

BAF170720000

Cleanware microscope optics cleaning kit

pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103200
NACRES:
NB.35

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F17072-0000

General description

This handy kit has everything you need, including ten pre-saturated optic and lens cleaning pads, two individually wrapped packs of anti-static plastic surface wipes and two 15.2 cm (6 in.) swabs which are pre-moistened with alcohol. Complete instructions for use included.

  • Includes 10 pre-saturated cleaning pads
  • Two packs of anti-static wipes

Legal Information

replacement part

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service