Skip to Content
Merck
All Photos(3)

Key Documents

B8681

Sigma-Aldrich

Benzoin

98%

Synonym(s):

α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone, 2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
391839
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

bp

194 °C/12 mmHg (lit.)

mp

134-138 °C (lit.)

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Benzoin is an industrially important chemical that is used as an intermediate in the synthesis of different chemical additives, dyestuff, pharmaceuticals and also as a photoinitiator.

Application

Benzoin may be used as a photoinitiator in the synthesis of biocompatible and degradable hydrogels based on 2-hydroxyethyl (meth)acrylates and gelatin via combined microwave-assisted and photo-polymerization technique.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alkaline modified g-C3N4 photocatalyst for high selective oxide coupling of benzyl alcohol to benzoin.
Sun X, et al.
Applied Catalysis. B, Environmental, 220, 553-560 (2018)
The innovative combined microwave-assisted and photo-polymerization technique for synthesis of the novel degradable hydroxyethyl (meth) acrylate/gelatin based scaffolds.
Babic MM, et al.
Materials Letters, 213, 236-240 (2018)
Structures and reactivities of O-methylated Breslow intermediates.
Biplab Maji et al.
Angewandte Chemie (International ed. in English), 51(41), 10408-10412 (2012-09-13)
Javier Izquierdo et al.
Angewandte Chemie (International ed. in English), 51(47), 11686-11698 (2012-10-18)
N-Heterocyclic carbene (NHC) catalyzed transformations have emerged as powerful tactics for the construction of complex molecules. Since Stetter's report in 1975 of the total synthesis of cis-jasmon and dihydrojasmon by using carbene catalysis, the use of NHCs in total synthesis
Qiu-Fen Hu et al.
Journal of natural products, 76(10), 1854-1859 (2013-09-26)
Eight new C-4-alkylated deoxybenzoins (1-8), three new diphenylethylenes (9-11), and five known diphenylethylenes were isolated from Arundina graminifolia. The structures of 1-11 were elucidated by spectroscopic methods including extensive 1D and 2D NMR techniques. Compounds 9-11 are the first naturally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service