Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
B25401
Benzyl mercaptan
99%
Synonym(s):
α-Toluenethiol, α-Tolyl mercaptan, Benzenemethanethiol
About This Item
Recommended Products
vapor density
>4 (vs air)
Assay
99%
form
liquid
refractive index
n20/D 1.575 (lit.)
bp
194-195 °C (lit.)
density
1.058 g/mL at 25 °C (lit.)
SMILES string
SCc1ccccc1
InChI
1S/C7H8S/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
InChI key
UENWRTRMUIOCKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- A nucleophilic reagent in the cleavage of proanthocyanidins into their constitutive subunits.[1]
- A reactant in the synthesis of dithiocarboxylic esters in the presence of phosphorus pentasulfide as a catalyst.[2]
- A modifier to functionalize the surface of CNT for enhanced interaction with Pt-nanoparticles.[3]
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
Do you have dibenzyl disulphide?
1 answer-
There are currently two product options for Dibenzyl disulfide, 33820 and B21805.
33820 Dibenzyl sulfide is has a minimum purity specification of 95%.
See the link below to review the product:
https://www.sigmaaldrich.com/US/en/product/aldrich/33820
See the link below to review a sample Certificate of Analysis:
https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/PDF331003.pdfB21805 Dibenzyl disulfide has a minimum purity specification of 98%.
See the link below to review the product page:
https://www.sigmaaldrich.com/US/en/product/aldrich/b21805
See the link below to review a sample Certificate of Analysis:
https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/B21805-BULKMKCR5263.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service