Skip to Content
Merck
All Photos(1)

Key Documents

ALD00558

Sigma-Aldrich

1-((3,5-Difluorophenyl)sulfonyl)bicyclo[1.1.0]butane

95%

Synonym(s):

Baran cyclobutylation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8F2O2S
Molecular Weight:
230.23
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation

storage temp.

2-8°C

General description

1-((3,5-Difluorophenyl)sulfonyl)bicyclo[1.1.0]butane can be used as a cyclobutylation agent to introduce bicyclobutane group onto a variety of amines and thiols without affecting other functional groups.

Application

The following "spring-loaded" electrophile is a convenient reagent for the installation of cyclobutane to primary and secondary amines. This product is one of several synthetically useful reagents reported by Baran and coworkers on strain-release amination.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

1-((3, 5-Difluorophenyl) sulfonyl) bicyclo [1.1. 0] butane
Lopchuk JM and Baran PS
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-3 (2001)

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service