Skip to Content
Merck
All Photos(1)

Documents

A86406

Sigma-Aldrich

α-Angelica lactone

98%

Synonym(s):

alpha-Angelica lactone, 4-Hydroxy-3-pentenoic acid γ-lactone, 5-Methyl-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
Beilstein:
108394
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.448 (lit.)

bp

55-56 °C/12 mmHg (lit.)

mp

13-17 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

SMILES string

CC1=CCC(=O)O1

InChI

1S/C5H6O2/c1-4-2-3-5(6)7-4/h2H,3H2,1H3

InChI key

QOTQFLOTGBBMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Huang et al.
Chemical communications (Cambridge, England), 48(18), 2439-2441 (2012-01-24)
The first organocatalytic asymmetric assembly of Morita-Baylis-Hillman carbonates of isatins and α-angelica lactone has been studied, affording multifunctional products containing two valuable pharmacophores and vicinal quaternary chiral centers in high stereoselectivity (up to 92% ee, dr >95:5).
Y M Ioannou et al.
Cancer research, 42(4), 1199-1204 (1982-04-01)
The effects of alpha-angelica lactone (alpha-AL), butylated hydroxyanisole (BHA), and beta-naphthoflavone (beta-NF) on the amount of benzo(alpha)pyrene (BP) metabolite:DNA adducts formed in the forestomach, lung, and liver of ICR/Ha mice were investigated 48 hr after p.o. administration of BP. BP
Lin Zhou et al.
Organic letters, 13(12), 3056-3059 (2011-05-21)
A direct highly diastereo- and enantioselective asymmetric vinylogous Mannich-type (AVM) reaction of aldimines with nonactivated natural α-angelica lactone has been successfully developed. It was demonstrated that the nonactivated natural α-angelica lactone is a useful vinylogous nucleophile to give the chiral
W A Nijhoff et al.
Carcinogenesis, 16(3), 607-612 (1995-03-01)
The naturally occurring anticarcinogens flavone and alpha-angelicalactone incorporated separately and simultaneously in the diet at 0.5, 0.1, 0.05 and 0.01% w/w, were studied with respect to their effects on oesophageal, gastric, intestinal, colonic and hepatic (i) glutathione S-transferase (GST) enzyme
Hagai Tavori et al.
Bioorganic & medicinal chemistry, 16(15), 7504-7509 (2008-06-24)
Paraoxonase1 (PON1) is a HDL bound enzyme and many of the anti-atherogenic properties of HDL are attributed to PON1. The enzyme precise mechanism of protective action and its endogenous substrate remain elusive. PON1 hydrolyzes organophosphates, arylesters and lactones, whereas the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service