Skip to Content
Merck
All Photos(2)

Key Documents

A805

Sigma-Aldrich

Acenaphthylene

75%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8
CAS Number:
Molecular Weight:
152.19
Beilstein:
774092
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

75%

form

solid

impurities

20% acenaphthene

bp

280 °C (lit.)

mp

78-82 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

c1cc2C=Cc3cccc(c1)c23

InChI

1S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8H

InChI key

HXGDTGSAIMULJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiru Ma et al.
The journal of physical chemistry. A, 111(10), 2020-2027 (2007-03-29)
13C chemical shift tensor data from 2D FIREMAT spectra are reported for 4,7-di-t-butylacenaphthene and 4,7-di-t-butylacenaphthylene. In addition, calculations of the chemical shielding tensors were completed at the B3LYP/6-311G** level of theory. While the experimental tensor data on 4,7-di-t-butylacenaphthylene are in
Onruthai Pinyakong et al.
FEMS microbiology letters, 238(2), 297-305 (2004-09-11)
Sphingomonas sp. strain A4 is capable of utilizing acenaphthene and acenaphthylene as sole carbon and energy sources, but it is unable to grow on other polycyclic aromatic hydrocarbons (PAHs). The genes encoding terminal oxygenase components of ring-hydroxylating dioxygenase (arhA1 and
N D Marsh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(8), 1499-1511 (2000-07-25)
We have examined the UV spectra of six newly-synthesized ethynyl-PAH (1-ethynylnaphthalene, 5-ethynylacenaphthylene, 1-ethynylanthracene, 9-ethynylphenanthrene, 3-ethynylfluoranthene, and 1-ethynylpyrene) and five corresponding CP-PAH (acenaphthylene, aceanthrylene, acephenanthrylene, cyclopenta[cd]fluoranthene, and cyclopenta[cd]pyrene) and have found the following systematic behavior: ethynyl-substitution results in average bathochromic shifts
Yan Hao et al.
Organic & biomolecular chemistry, 10(4), 724-728 (2011-12-15)
A simple and novel protocol for the efficient synthesis of a series of 8-carboxylnaphthyl functionalized pyrazolo[3,4-b]pyridine derivatives was developed through a one-pot, three-component reaction involving acenaphthylene-1,2-dione and 1H-pyrazol-5-amine in acetic acid medium. The reaction represents the first facile conversion of
E Leonhardt et al.
Analytical chemistry, 70(6), 1228-1230 (1998-04-08)
Polycyclic aromatic hydrocarbons were extracted from a soil sample using ultrasound and dichloromethane-, cyclohexane-, and toluene-water mixtures. It was found that when dichloromethane is used as an extractant, acenaphthylene reacts with the solvent. Several chlorinated and oxygenated derivatives were identified.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service