Skip to Content
Merck
All Photos(3)

Documents

A65182

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

~5% Water, technical grade, 95%

Synonym(s):

β-Aminoisobutyl alcohol, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
505979
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

3 (vs air)

vapor pressure

<1 mmHg ( 25 °C)

Assay

95%

form

liquid

impurities

≤5% 2-(Methylamino)-2-methyl-1-propanol
~5% Water

refractive index

n20/D 1.4455 (lit.)

useful pH range

9.0-10.5

pKa (25 °C)

9.7

bp

165 °C (lit.)

mp

24-28 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

SMILES string

CC(C)(N)CO

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in an efficient synthesis of 2-oxazolidinones via carbonylation with CO in the presence of salen-cobalt catalysts.
Used to derivatize carboxylic acids for GC analysis and to synthesize 2-oxazolines for further transformations.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 1010-1010 (1992)
Journal of Chromatography A, 673, 101-101 (1994)
Erik C B Johnson et al.
Tetrahedron letters, 48(10), 1795-1799 (2007-03-05)
Modification of a peptide-(α)thioester with a sequence of six arginines on the thioester leaving group can render soluble all peptides derived from a polytopic integral membrane protein. This strategy greatly simplifies the synthesis of peptide-(α)thioester building blocks for the total
Tetrahedron, 49, 9353-9353 (1993)
Shia-Chung Chen et al.
Journal of hazardous materials, 179(1-3), 692-700 (2010-04-15)
In this study, the recovery of carbon dioxide using an absorbent composed of 2-amino-2-methyl-l-propanol (AMP)+monoethanolamine (MEA)+piperazine (PZ) in polytetrafluoroethylene (PTFE) membrane contactors was investigated. Experiments were conducted using various gas flow rates, liquid flow rates, absorbent blends, and pore size

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service