A55306
2-(2-Pyridyl)ethylamine
95%
Synonym(s):
2-(2-Aminoethyl)pyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Beilstein:
111208
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.536 (lit.)
bp
92-93 °C/12 mmHg (lit.)
density
1.021 g/mL at 25 °C (lit.)
SMILES string
NCCc1ccccn1
InChI
1S/C7H10N2/c8-5-4-7-3-1-2-6-9-7/h1-3,6H,4-5,8H2
InChI key
XPQIPUZPSLAZDV-UHFFFAOYSA-N
Gene Information
human ... HRH1(3269)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Susanne Mommert et al.
British journal of pharmacology, 177(3), 600-613 (2019-07-23)
Human monocyte-derived M1 macrophages develop in relation to growth factors, bacterial products, and cytokines in a local micro-environment. M1 macrophages produce pro-inflammatory mediators, in particular, oncostatin M (OSM), which is secreted from the cells in response to the active complement
H C Liang et al.
Inorganic chemistry, 39(26), 5884-5894 (2001-02-24)
Copper-dioxygen interactions are of interest due to their importance in biological systems as reversible O2- carriers, oxygenases, or oxidases and also because of their role in industrial and laboratory oxidation processes. Here we report on the kinetics (stopped-flow, -90 to
P K Rangachari et al.
The Journal of pharmacology and experimental therapeutics, 271(2), 1016-1026 (1994-11-01)
The receptors involved in the neural and non-neural effects of histamine on the canine proximal colon in vitro were defined functionally by using selective agonists and antagonists. Two preparations, a ganglionated mucosa and an aganglionated epithelium, were set up in
C H Gelband et al.
Circulation research, 73(1), 24-34 (1993-07-01)
The patch-clamp technique was used to examine the sensitivity of delayed rectifier K+ channels to changes in intracellular divalent cations (Mg2+ and Ca2+). During voltage-step and ramp depolarizations, a delayed rectifier K+ current (IK(dr)) was identified in renal, pulmonary, coronary
Katrin Schaper-Gerhardt et al.
British journal of pharmacology, 177(3), 614-622 (2018-12-28)
Th9 cells represent a recently defined subset of CD4+ T-helper cells, characterized by a high production of IL-9. They are found at increased frequency in lesions of atopic dermatitis, where IL-9 is also elevated. As histamine is up-regulated in lesions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service