Skip to Content
Merck
All Photos(1)

Documents

938386

Sigma-Aldrich

Barium

new

beads, ≤2.0 mm particle size, 99.99% trace metals basis

Synonym(s):

Elemental Barium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ba
CAS Number:
Molecular Weight:
137.33
MDL number:
UNSPSC Code:
12141505

Quality Level

Assay

99.99% trace metals basis

form

beads

resistivity

50.0 μΩ-cm, 20°C

particle size

≤2.0 mm

bp

1640 °C (lit.)

mp

725 °C (lit.)

density

3.6 g/mL at 25 °C (lit.)

SMILES string

[Ba]

InChI

1S/Ba

InChI key

DSAJWYNOEDNPEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barium is a silvery white soft alkaline earth metals. It easily oxidizes in air and water. The electrical conductivity of Barium is very high. Barium is even more chemically reactive than Magnesium, calcium and strontium. It is an essential component of high temperature superconductors.

Application

Critical for the synthesis of pure Ba21M2O5HX (M = Ge, SI, Ga, In, Tl) suboxide Zintl phases.3
In the semiconductor and electronics industries, thinner coatings are required due to newer technology and design. Therefore, it is crucial to reduce unwanted metals or interfering elements to achieve the intended performances. Our Barium beads, which have a 99.99% trace metal basis, are the ideal choice when it comes to high-end thin film coating materials. The undesirable metals have been measured to the ppm level using ICP-Ms. Elemental Barium has wide applications. For example: In order to create barium doped oxide with high carrier mobility and improved electrical stability for thin film transition, barium is utilized in a low temperature solution process

Furthermore, metallic barium is used to produce Barium-Containing p-tert-Butylcalix[4]arene, which is a precursor for the fabrication of BaOTiO2 thin films via dip coating on a sapphire substrate It can be used as a dopant for Pb2+replacement in perovskite films. The addition of barium enhances the power conversion efficiency of organic-inorganic lead halide solar cells.

Interestingly, the solid-state synthesis method is used to synthesize quaternary chalcogenides with thermolectric properties using metallic Ba

Furthermore, barium metal is also used in the highly selective and stereo controlled homocoupling reaction of allylic halide

Our barium beads 0.5-2.0 mm particle size, 99.99% trace metals basis will be ideal precursor materials for the diverse applications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of five new layered quaternary chalcogenides SrScCuSe3, SrScCuTe3, BaScCuSe3, BaScCuTe3, and BaScAgTe3: crystal structures, thermoelectric properties, and electronic structures
Ishtiyak M, et al.
Inorganic chemistry frontiers, 8, 4086-4101 (2021)
High Performance, Low Temperature Solution-Processed Barium and Strontium Doped Oxide Thin Film Transistors
Banger K K, et al.
Chemistry of Materials, 26(2), 1195-1203 (2014)
New Calix[4]arene-Based Barium Precursor for BaO?TiO2Thin Film Deposition
Burtman V, et al.
Chemistry of Materials, 9, 3101?3106-3101?3106 (1997)
Highly selective homocoupling reaction of allylic halides using barium metal
Yanagisawa A, et al.
The Journal of Organic Chemistry, 57, 6386?6387-6386?6387 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service