Skip to Content
Merck
All Photos(1)

Key Documents

935522

Sigma-Aldrich

Palladium(II) sulfate dihydrate

≥99.9% trace metals basis

Synonym(s):

Palladium monosulfate dihydrate, Palladium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PdSO4 · 2H2O
UNSPSC Code:
12352311
NACRES:
NA.21
Assay:
≥99.9% trace metals basis
43.2-46.0% Pd basis (gravimetric)
form:
powder or crystals (or Chunk(s))
solubility:
water: soluble

Quality Level

Assay

≥99.9% trace metals basis
43.2-46.0% Pd basis (gravimetric)

form

powder or crystals (or Chunk(s))

impurities

≤1000 ppm (trace metals analysis)

color

dark red to brown

pH

1-9

solubility

water: soluble

Related Categories

General description

Palladium Sulphate dihydrate is a brown to red crystalline powder that is water soluble. It formed through the interaction of palladium metal and sulfuric acid.

Application

In general, palladium(II) sulphate dihydrate is used to produce catalysts, Pd nanoparticles, and Pd complexes with potential applications in catalysis.For example: Alumina was used as a support material and PdSO4 as a palladium precursor to manufacture 4% PdSO4/Al2O3 catalyst for current lean-burn methane combustion for automotive applications Furthermore, Pd(0) atoms are produced by catalysing the semi-hydrogenation reaction with remarkably high efficiency utilising PdSO4 salts with H2 in alcohol solutions, regardless of the alkyne

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parts-Per-Million of Soluble Pd0 Catalyze the Semi-Hydrogenation Reaction of Alkynes to Alkenes
Ballesteros-Soberanas J, et al.
The Journal of Organic Chemistry, 88, 18-26 (2023)
Case study of a modern lean-burn methane combustion catalyst for automotive applications: What are the deactivation and regeneration mechanisms?
Kinnunen N M, et al.
Applied Catalysis. B, Environmental, 207, 114-119 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service