Skip to Content
Merck
All Photos(2)

Key Documents

935123

Sigma-Aldrich

Infrared Ag2S quantum dots

λmax, 800 nm, 10 mg/mL in DMF

Synonym(s):

Fluorescent nanocrystals, INFIQ® quantum dots, Silver based core type QDs

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2S
Molecular Weight:
247.80
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

description

>= 4 (peak/valley ratio)

Quality Level

form

liquid

concentration

10 mg/mL in DMF

diameter

<15 nm

λmax

≤70 nm (FWHM)
800 nm

storage temp.

2-8°C

General description

Silver sulfide (Ag2S) quantum dots (QD) are semiconducting inorganic nanoparticles, offered as a concentrated solution (10 mg/mL) in N,N-dimethylformamide. The most important property of a Ag2S QD is its excitonic absorption peak wavelength, which is controlled by the particle size. These infrared Ag2S quantum dots have an excitonic absorption peak wavelength of 800 nm and narrow peak width, indicating that they are a monodisperse ensemble of nanoparticles, all a very similar size. In addition, Ag2S quantum dots do not contain toxic elements (such as Cd and Pb), making them a greener alternative to Cd- and Pb-based QDs.

Application

Ag2S quantum dots have a range of potential applications in nanodiagnostics, solar cells, and photoelectrochemical biosensors. These nanocrystals can be used as NIR optical imaging probes in research activities aimed at studying nanodiagnostics and imaging in vivo. Additionally, studies have shown that Ag2S QDs can serve as a highly efficient and broadband sensitizer for solar cells, making them a promising material for research and development activities in the field of photovoltaics Ag2S QDs have also demonstrated potential as PEC biosensors, with studies showing that they have extensive potential in this application due to their simplicity, low toxicity, sensitivity, and wide range of potential uses

Legal Information

INFIQ is a registered trademark of Quantum Science Ltd.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-toxic Ag2S quantum dots for photoelectrochemical detection glucose and cancer cells
Zhang X, et al.
Biosensors And Bioelectronics, 56, 307-312 (2014)
Ag2S quantum dot-sensitized solar cells
Tubtimtae A, et al.
Electrochemistry Communications, 12, 1158-1160 (2010)
Peng Jiang et al.
Biomaterials, 33(20), 5130-5135 (2012-04-10)
A one-step method for synthesizing water-soluble Ag(2)S quantum dots terminated with carboxylic acid group has been reported. The crystal structure and surface of the prepared Ag(2)S quantum dots were characterized. The prepared Ag(2)S quantum dots exhibited bright photoluminescence and excellent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service