Skip to Content
Merck
All Photos(1)

Key Documents

934070

Sigma-Aldrich

Ovine collagen

3 mg/ml solution, low endotoxin

Synonym(s):

CollOvine RD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.28

Quality Level

form

liquid

impurities

low endotoxin
≤100 EU/g Endotoxin (collagen)

pH

2.5-4

density

1.01 g/mL± 0.02 g/mL

General description

Collagen is a major component of the extracellular matrix (ECM) and offers low immunogenicity, a porous structure, good permeability, and biocompatibility to promote cell viability, adhesion, spreading, proliferation and differentiation. Low endotoxin ovine collagen solution is derived from sheep and is a bovine or porcine alternative that is highly purified, fully traceable, and low endotoxin making it suitable for many biomedical applications.

Application

  • 3D bioprinting
  • Tissue engineering
  • Drug delivery
  • Regenerative medicine
  • Cell culture, encapsulation, expansion and differentiation

Features and Benefits

  • Certified Disease Free Sheep
  • 93-97% Type I Collagen
  • 3-7% Type III Collagen
  • 99% Purity
  • Prion Free

Legal Information

CollOvine is a trademark of OviGenex LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Egor Olegovich Osidak et al.
International journal of bioprinting, 6(3), 270-270 (2020-10-23)
Biomaterials made using collagen are successfully used as a three-dimensional (3D) substrate for cell culture and considered to be promising scaffolds for creating artificial tissues. An important task that arises for engineering such materials is the simulation of physical and
Elizabeth E Antoine et al.
Tissue engineering. Part B, Reviews, 20(6), 683-696 (2014-06-14)
Type I collagen hydrogels have been used successfully as three-dimensional substrates for cell culture and have shown promise as scaffolds for engineered tissues and tumors. A critical step in the development of collagen hydrogels as viable tissue mimics is quantitative
Atiqah Salleh et al.
Biomedicines, 10(4), 816-816 (2022-04-24)
Tissue engineering products have grown rapidly as an alternative solution available for chronic wound and burn treatment. However, some drawbacks include additional procedures and a lack of antibacterial properties that can impair wound healing, which are issues that need to
Jana Stepanovska et al.
Biomedicines, 9(9), 1137-1137 (2021-09-29)
Bioprinting is a modern tool suitable for creating cell scaffolds and tissue or organ carriers from polymers that mimic tissue properties and create a natural environment for cell development. A wide range of polymers, both natural and synthetic, are used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service