Skip to Content
Merck
All Photos(2)

Documents

932515

Sigma-Aldrich

Copper(II) phthalocyanine

≥98%

Synonym(s):

CuPc, Phthalocyanine blue, Pigment Blue 15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16CuN8
CAS Number:
Molecular Weight:
576.07
Colour Index Number:
74160
Beilstein:
4121848
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

grade

sublimed grade

Quality Level

description

µh ≈ 2.8 x 10-4 cm2V-1s-1

Assay

≥98%

loss

0.5% TGA, >430ºC (weight loss)

mp

350 °C

solubility

DMF: soluble
dichloromethane: soluble

fluorescence

λem 404 nm in dichloromethane (PL in film)

Orbital energy

HOMO 5.2 eV 
LUMO 3.5 eV 

SMILES string

c1ccc2c(c1)C3=NC4=[N@@H]5C(=Nc6n7c(N=C8c9ccccc9C%10=[N@@H]8[Cu]57N3C2=N%10)c%11ccccc6%11)c%12ccccc4%12

InChI

1S/C32H16N8.Cu/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

XCJYREBRNVKWGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) phthalocyanine (CuPC) find uses in high-efficiency red electrophosphorescent OLEDs, transistors and OPV
Copper(II) phthalocyanine can be used as an electron donor material or be incorporated into the active layer of organic photovoltaic devices (OPVs). Since it exhibits excellent charge transport properties, it can be be utilized as a p-type organic semiconductor in studies to improve device performance and electrical behavior of Organic Field-Effect Transistors (OFETs). It can be employed as an electron-blocking or hole-blocking layer in organic light-emitting diodes(OLEDs) devices for studies to improve efficiency and device stability. It has been used to conduct research in chemical sensing applications, and organic thin-film transistors (OTFTs).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronic structure and bonding in metal phthalocyanines, Metal=Fe, Co, Ni, Cu, Zn, Mg
Liao, Meng-Sheng; et al.
Journal of Chemical Physics, 114, 9780-9791 (2001)
Influence of the metal phthalocyanine molecular orientation on charge separation at the organic donor/acceptor interface
Lim, Heeseon
Journal of Material Chemistry C, 9, 2156-2164 (2021)
Functional supramolecular gels based on the hierarchical assembly of porphyrins and phthalocyanines
Feng, Xuenan, et al.
Frontiers in Chemistry, 7 (2019)
Obtaining high-efficiency red electrophosphorescent OLEDs by changing the thickness of light-emitting layer
Zhang, et al.
Displays, 28, 150?153-150?153 (2007)
Functional supramolecular gels based on the hierarchical assembly of porphyrins and phthalocyanines
Feng, Xuenan; et al.
Frontiers in Chemistry, 7 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service