Skip to Content
Merck
All Photos(2)

Key Documents

932507

Sigma-Aldrich

N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)-2,2′-dimethylbenzidine

≥99% (HPLC)

Synonym(s):

2,2′-Dimethyl-N,N′-di-[(1-naphthyl)-N,N′-diphenyl]-1,1′-biphenyl-4,4′-diamine, α-NPD, N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)-2,2′-dimethylbenzidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H36N2
CAS Number:
Molecular Weight:
616.79
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

loss

0.5% TGA, > 310 °C (weight loss)

solubility

THF: soluble
chlorobenzene: soluble
chloroform: soluble

fluorescence

λem 447 nm±20 nm in THF

Orbital energy

HOMO 5.4 eV 
LUMO 2.3 eV 

λ

in THF

UV absorption

λ: 307 nm±5 nm Amax

SMILES string

Cc1cc(ccc1-c2ccc(cc2C)N(c3ccccc3)c4cccc5ccccc45)N(c6ccccc6)c7cccc8ccccc78

InChI

1S/C46H36N2/c1-33-31-39(47(37-19-5-3-6-20-37)45-25-13-17-35-15-9-11-23-43(35)45)27-29-41(33)42-30-28-40(32-34(42)2)48(38-21-7-4-8-22-38)46-26-14-18-36-16-10-12-24-44(36)46/h3-32H,1-2H3

InChI key

ZJFKMIYGRJGWIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hole Transport/Electron Blocking Layer (HTL/EBL) for Efficient Organic Light Emitting Diode (OLED).
N,N′-Bis(naphthalen-1-yl)-N,N′-bis(phenyl)-2,2′-dimethylbenzidine can be used in organic light-emitting diodes (OLEDs) as a hole transport material. It has a suitable energy level and molecular structure that allows for effective energy transfer to dopant molecules, leading to enhanced electroluminescence and color purity in OLED devices. Due to its excellent charge transport properties and sensitivity to light, it can be used in studies such as organic photodetectors. It can be used as an active layer or part of the photodetection device structure, enabling efficient charge generation and transport upon light exposure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Efficient, Red Delayed Fluorescent Emitters with Exothermic Reverse Intersystem Crossing via Hot Excited Triplet States.
Wang R, et al.
The Journal of Physical Chemistry C, 124, 20816?20826-20816?20826 (2020)
Impedance Characterization of Organic Light-Emitting Structures with Thermally Activated Delayed Fluorescence.
Voitsekhovskii A V, et al.
Physica Status Solidi RRL: Rapid Research Letters, 217, 1900847-1900847 (2020)
Dual-acceptor thermally activated delayed fluorescence emitters: Achieving high efficiency and long lifetime in orange-red OLEDs.
Hu X, et al.
Chemical Engineering Journal, 434, 134728-134728 (2022)
Highly efficient white light-emitting diodes with a bi-component emitting layer based on blue and yellow thermally activated delayed fluorescence emitters.
Chen Y, et al.
Journal of Material Chemistry C, 6, 2951-2956 (2018)
Effect of Triplet Confinement on Triplet-Triplet Annihilation in Organic Phosphorescent Host-Guest Systems.
Ligthart A, et al.
Advanced Functional Materials , 28, 1804618-1804618 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service