Skip to Content
Merck
All Photos(1)

Documents

931594

Sigma-Aldrich

Pomalidomide-PEG3-OH

≥95%

Synonym(s):

1H-Isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-4-[[2-[2-(2-hydroxyethoxy)ethoxy]ethyl]amino], 2-(2,6-Dioxo-3-piperidinyl)-4-[[2-[2-(2-hydroxyethoxy)ethoxy]ethyl]amino]-1H-isoindole-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O7
CAS Number:
Molecular Weight:
405.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

ligand

pomalidomide

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

Application

Pomalidomide-PEG3-OH enables the synthesis of molecules for degradation of proteins and PROTAC® (proteolysis-targeting chimeras) research. This conjugate contains a Cereblon (CRBN) recruiting ligand, a PEG linker, and a pendant hydroxyl group. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks, parallel synthesis can be used to more quickly generate degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.


Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation

Protein Degrader Building Blocks

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service