Skip to Content
Merck
All Photos(1)

Key Documents

928003

Sigma-Aldrich

Ready-to-Cast LiNi0.33Mn0.33Co0.33O2 (NMC111) Slurry for Lithium ion battery

Synonym(s):

Blade coatable ink slurry for Lithium ion battery, NMC, NMC111, NMC111 ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

type

(cathode materials)

form

paste

composition

LiNi0.33Mn0.33Co0.33O2
>40 wt% active material

45-60 wt% solid content

impurities

<0.5% H2O

application(s)

battery manufacturing

General description

Our ready-to-cast NMC111 slurry is a liquid-type dispersion, made up of battery-grade lithium-nickel-manganese-cobalt-oxide active material, conductive additives, binder, and organic solvent. The product features a low moisture content, which is key for fabricating smooth pinhole-free cathodes.

Application

Our ready-to-cast NMC111 slurry is designed to be used for casting films of NMC111 cathodes for secondary lithium-ion batteries and can be used directly for blade-coating or slot-die coating onto aluminum foil current collectors. Resulting from precise formulation, our ready-to-cast NMC111 slurry is stable and processable and easy to form defect-free films of the nickel-rich NMC111 cathode material.
The liquid-type dispersion is made up of a common active material, a conductive agent, a binder and a solvent that researchers can readily use for processing lithium-ion battery electrodes.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.0 °F

Flash Point(C)

32.77 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service