Skip to Content
Merck
All Photos(1)

Key Documents

920924

Sigma-Aldrich

Monolayer hexagonal Boron Nitride (hBN) on Si/SiO2 wafer

diam. 200 mm (8 in.)

Synonym(s):

Boronitrene, Hexagonal boron nitride monolayer, Single-layer hexagonal boron nitride, White graphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Pricing and availability is not currently available.

description

Bandgap: 5.97 eV
Metal Impurities: 1.00e10 – 5.00e10 (at/cm2)
Monolayer h-BN
Orientation: <1-0-0>
Raman Peak: 1370 /cm-1
Substrate Type/Doping: P/B
hBN Coverage: 100% with sporadic adlayers

Quality Level

resistivity

5-30 Ω-cm

diam.

200 mm (8 in.)

thickness

300 nm , Oxide
725 ± 25 μm , Wafer

grain size

>4 μm

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monolayer hexagonal boron nitride (h-BN), also known as “white graphene”, is a wide-bandgap 2D crystal (~6 eV that can be tuned to ~2 eV) [1][2] with exceptional strength [3], large oxidation resistance at high temperatures [3], and optical functionalities [2][3]. Among its potential applications are:
  • Two-dimensional electronics [1][2][3]
  • Nanophotonic and other optoelectronic devices [1][2][3]
  • Quantum communication and information science [2]
  • Aerospace industry [4]
  • MEMS and NEMS [5]
  • Micro-/nano- actuators [3]
  • nsulating/transparent coatings [1]

Storage and Stability

To ensure the maximum shelf life of your hBN sample, it is best stored under vacuum or in inert atmosphere (Argon or Nitrogen) conditions once the vacuum sealed package has been opened.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical and Bandgap Engineering in Monolayer Hexagonal Boron Nitride
Ba K, et al.
Scientific Reports, 7 (2017)
Monolayer to Bulk Properties of Hexagonal Boron Nitride
Wickramaratne D, et al.
Physical Chemistry, 122, 25524 25529-25524 25529 (2018)
Hexagonal boron nitride nanomechanical resonators with spatially visualized motion
Zheng X, et al
Microsystems & Nanoengineering (2017)
Piezoelectricity in Monolayer Hexagonal Boron Nitride
Ares P, et al.
Advanced Materials, 32 (2020)
From 2-D to 0-D Boron Nitride Materials, The Next Challenge
Stagi L, et al.
Materials, 12(23) (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service