Skip to Content
Merck
All Photos(2)

Key Documents

914827

Sigma-Aldrich

N6-((Prop-2-yn-1-yloxy)carbonyl)-L-lysine hydrochloride

≥98%

Synonym(s):

(S)-Amino-6-((prop-2-ynyloxy)carbonylamino)hexanoic acid hydrochloride, Clickable amino acid for bioconjugation, H-L-Lys(Poc)-OH HCl, Lysine-alkyne, Plk, Propargyl-derivatized lysize, UAA crosslinker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O4 · xHCl
CAS Number:
Molecular Weight:
228.25 (free base basis)
UNSPSC Code:
12352200

Assay

≥98%

form

powder

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Schmidt et al.
ACS chemical biology, 13(9), 2472-2483 (2018-08-01)
Single-molecule techniques allow unique insights into biological systems as they provide unrivaled access to structural dynamics and conformational heterogeneity. One major bottleneck for reliable single-molecule Förster resonance energy transfer (smFRET) analysis is the identification of suitable fluorophore labeling sites that
Yiming Li et al.
Organic & biomolecular chemistry, 11(16), 2624-2629 (2013-03-02)
Three alkyne-containing pyrrolysine derivatives were synthesized and genetically encoded into proteins by a mutant PylRS-tRNA pair with high efficiencies. With these alkyne handles, site-specific dual labeling of proteins can be achieved via a bioorthogonal thiol-yne ligation reaction.
Claudio Zambaldo et al.
Journal of the American Chemical Society, 139(34), 11646-11649 (2017-08-16)
Nisin is a complex lanthipeptide that has broad spectrum antibacterial activity. In efforts to broaden the structural diversity of this ribosomally synthesized lantibiotic, we now report the recombinant expression of Nisin variants that incorporate noncanonical amino acids (ncAAs) at discrete
Ivana Nikić et al.
Angewandte Chemie (International ed. in English), 53(8), 2245-2249 (2014-01-30)
The growing demands of advanced fluorescence and super-resolution microscopy benefit from the development of small and highly photostable fluorescent probes. Techniques developed to expand the genetic code permit the residue-specific encoding of unnatural amino acids (UAAs) armed with novel clickable
K W Swiderska et al.
Bioorganic & medicinal chemistry, 25(14), 3685-3693 (2017-05-20)
Recent advances in site-specific protein modification include the increasingly popular incorporation of unnatural amino acid(s) using amber codon, a method developed by Schultz and coworkers. In this study, we employ this technique to introduce propargyllysine (PrK) in human fibroblast growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service