Skip to Content
Merck
All Photos(1)

Key Documents

912069

Sigma-Aldrich

Morpholinium bromide

Synonym(s):

MORPHB, Morpholine hydrobromide (8CI,9CI), Morpholine monohydrobromide, Morpholinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10BrNO
CAS Number:
Molecular Weight:
168.03
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

description

Identity by (1H NMR)

Quality Level

form

powder

color

white

InChI

1S/C4H9NO.BrH/c1-3-6-4-2-5-1;/h5H,1-4H2;1H

InChI key

BVJOXYJFOYNQRB-UHFFFAOYSA-N

Application

Organohalide based perovskites have emerged as an important class of material for solar cell applications. The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells.

Storage and Stability

Store in dark at 20 °C

Legal Information

Product of Greatcell Solar®
Greatcell Solar is a registered trademark of Greatcell Solar

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Efficiency and High Open Circuit Voltage in Quasi 2D Perovskite Based Solar Cells.
Cohen B E, et al.
Advances in Functional Materials (2016)
Shanshan Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 31(30), e1901090-e1901090 (2019-06-06)
2D Ruddlesden-Popper perovskite (RPP) solar cells have excellent environmental stability. However, the power conversion efficiency (PCE) of RPP cells remains inferior to 3D perovskite-based cells. Herein, 2D (CH3 (CH2 )3 NH3 )2 (CH3 NH3 )n-1 Pbn I3n+1 perovskite cells with
Lingling Mao et al.
Journal of the American Chemical Society, 139(14), 5210-5215 (2017-03-18)
Hybrid inorganic-organic perovskites are developing rapidly as high performance semiconductors. Recently, two-dimensional (2D) perovskites were found to have white-light, broadband emission in the visible range that was attributed mainly to the role of self-trapped excitons (STEs). Here, we describe three
Eran Edri et al.
The journal of physical chemistry letters, 4(6), 897-902 (2013-03-21)
Mesoscopic solar cells, based on solution-processed organic-inorganic perovskite absorbers, are a promising avenue for converting solar to electrical energy. We used solution-processed organic-inorganic lead halide perovskite absorbers, in conjunction with organic hole conductors, to form high voltage solar cells. There

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service