Skip to Content
Merck
All Photos(2)

Key Documents

906034

Sigma-Aldrich

Ammonium pentaborate octahydrate

Synonym(s):

Ammonium boron oxide octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)B5O8 · 8H2O
CAS Number:
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23
Assay:
≥99.0% (with H2SO4, titration)
form:
powder

Assay

≥99.0% (with H2SO4, titration)

form

powder

InChI

1S/B.H3N.H2O.O/h;1H3;1H2;/p+1

InChI key

CUSUZJNFWWWYIV-UHFFFAOYSA-O

Application

Ammonium pentaborate is an alkaline salt utilized in the creation of thin films by developing an oxide film which can inhibit corrosion of metals. It finds applications in the preparation of electrolytic capacitors, as a solvent for metallic oxides, as a flame retardant additive, in synthesis of nitrogen-doped ZnO particles, and in treatment of carbon nanotmaterials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impedance and Corrosion Resistance Characteristics of Reanodized Anodic Alumina Film on AA 5052
N. Verma, et al.
Der Pharma Chemica, 10, 4-4 (2018)
T Balakrishnan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 578-583 (2008-03-11)
Nonlinear optical single crystals of ammonium pentaborate (APB) were grown by the slow cooling method from aqueous solution. Grown crystal was characterized by powder X-ray diffraction (PXRD) and FT-IR spectral analysis. Perfection of the grown crystal was evaluated by high-resolution
Polyamide 66/Nanoclay Composites: Synthesis, Thermal And Flammability Properties
S. Rathi, J.B. Dahiya
Advanced Materials Letters, 3, 5-5 (2012)
Fabrication of precision integrated capacitors
J. Jiang, P. Kohl
Thin Solid Films, 634-634 (2017)
Hongqi Sun et al.
Chemical communications (Cambridge, England), 49(85), 9914-9916 (2013-08-16)
Nitrogen (5.61 at%) doped reduced graphene oxide synthesized via a facile method was demonstrated as a superior metal-free catalyst for activation of peroxymonosulfate. Codoping with boron would further enhance the catalytic activity and the stability, providing a promising green material

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service