902586
Hydroxy-PEG4-t-butyl ester
Synonym(s):
tert-Butyl-1-hydroxy-3,6,9,12-tetraoxapentadecan-15-oate, HO-PEG4-CO-OtBu
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H30O7
CAS Number:
Molecular Weight:
322.39
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
liquid
reaction suitability
reagent type: cross-linking reagent
refractive index
n/D 1.4492
density
1.04746 g/mL
functional group
ester
hydroxyl
storage temp.
2-8°C
SMILES string
O=C(OC(C)(C)C)CCOCCOCCOCCOCCO
InChI
1S/C15H30O7/c1-15(2,3)22-14(17)4-6-18-8-10-20-12-13-21-11-9-19-7-5-16/h16H,4-13H2,1-3H3
InChI key
FJRDXEGYAVAMLB-UHFFFAOYSA-N
Related Categories
Application
This heterobifunctional, PEGylated crosslinker features a hydroxyl group at one end and t-butyl-protected carboxylic acid at the other, which can be deprotected with acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. Hydroxy-PEG4-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.
Other Notes
Legal Information
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Proline-functionalized magnetic core-shell nanoparticles as efficient and recyclable organocatalysts for aldol reactions.
Yacob Z, et al
Advanced Synthesis & Catalysis, 354(17), 3259-3264 (2012)
A new route for the synthesis of 1-amino-3,6,9,12-?tetraoxapentadecan-15-oic acid.
Wu X, et al.
J. Chem. Res. (M), 40(6), 368-370 (2016)
Recruiting cytotoxic T cells to folate-receptor-positive cancer cells.
Sumith A Kularatne et al.
Angewandte Chemie (International ed. in English), 52(46), 12101-12104 (2014-02-28)
Sara V Orski et al.
Journal of the American Chemical Society, 132(32), 11024-11026 (2010-08-12)
Surfaces containing reactive ester polymer brushes were functionalized with cyclopropenone-masked dibenzocyclooctynes for the light activated immobilization of azides using catalyst-free click chemistry. The photodecarbonylation reaction in the amorphous brush layer is first order for the first 45 s with a
Efficient Synthesis of Polyethylene Glycol Mono-Carboxylate via Michael Conjugate Addition.
Chen Q. et al.
Synthetic Communications, 34(13) (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service