Skip to Content
Merck
All Photos(1)

Documents

90218

Sigma-Aldrich

tert-Butyl 3-hydroxypropionate

≥98.0% (GC)

Synonym(s):

1,1-Dimethylethyl 3-hydroxypropanoate, 3-Hydroxypropanoic acid tert-butyl ester, 3-Hydroxypropionate tert-butyl ester, 3-Hydroxypropionic acid tert-butyl ester, tert-Butyl 3-hydroxypropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2COOC(CH3)3
CAS Number:
Molecular Weight:
146.18
Beilstein:
1854888
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.424

density

0.990 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)OC(=O)CCO

InChI

1S/C7H14O3/c1-7(2,3)10-6(9)4-5-8/h8H,4-5H2,1-3H3

InChI key

NSXCURRVJMPAPA-UHFFFAOYSA-N

Application

tert-Butyl 3-hydroxypropionate is a key starting material for the synthesis of wortmannilactone C. It can also be used to prepare three-dimensional metal-fullerene frameworks (MFFs).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Three?Dimensional Metal?Fullerene Frameworks.
Kraft A, et al.
Chemistry?A European Journal , 22(17), 5982-5987 (2016)
Synthetic Approach to Wortmannilactone C.
Brandt D, et al.
Organic Letters, 17(4), 816-819 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service