Skip to Content
Merck
All Photos(1)

Documents

900727

Sigma-Aldrich

PbS core-type quantum dots

fluorescence λem 1600 nm, 10 mg/mL in toluene

Synonym(s):

Fluorescent nanocrystals, Lead sulfide, PbS QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbS
CAS Number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

liquid

Quality Level

concentration

10 mg/mL in toluene

fluorescence

λem 1600 nm

storage temp.

2-8°C

InChI

1S/Pb.H2S/h;1H2

InChI key

MIXDRAMRMDOQJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The size for our lead sulfide (PbS) quantum dots (QDs) varies between 2.5 to 8 nm and depending upon this, these QDs emit between 900-1600 nm. Our PbS QDs possess high quantum yield, sharp emission and exhibit narrow fluorescence band (full width at half maximum <100 nm), which make them suitable as light absorber or IR emitter in applications in solar cells, photodetectors and infrared light emitting diodes (LEDs).

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

45.5 °F - closed cup

Flash Point(C)

7.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient quantum dot near-infrared light-emitting diodes.
Gong X, et al.
Nature Photonics, 10, 253-257 (2016)
Xinzheng Lan et al.
Advanced materials (Deerfield Beach, Fla.), 28(2), 299-304 (2015-11-19)
A solution-based passivation scheme is developed featuring the use of molecular iodine and PbS colloidal quantum dots (CQDs). The improved passivation translates into a longer carrier diffusion length in the solid film. This allows thicker solar-cell devices to be built
Gerasimos Konstantatos et al.
Nature nanotechnology, 5(6), 391-400 (2010-05-18)
The detection of photons underpins imaging, spectroscopy, fibre-optic communications and time-gated distance measurements. Nanostructured materials are attractive for detection applications because they can be integrated with conventional silicon electronics and flexible, large-area substrates, and can be processed from the solution

Articles

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

In this article, the properties of some of the new non-cadmium based QDs along with different applications of QDs are summarized.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service