900568
2-Ketoglutaric acid-13C5
≥99 atom % 13C, ≥95% (CP)
Synonym(s):
2-Ketopentanedioic acid-13C5, 2-Oxoglutaric acid-13C5, 2-Oxopentanedioic acid-13C5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
13C5H6O5
CAS Number:
Molecular Weight:
151.06
MDL number:
UNSPSC Code:
12352106
Recommended Products
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥95% (CP)
form
solid
technique(s)
bio NMR: suitable
mass shift
M+5
storage temp.
2-8°C
SMILES string
O[13C](=O)[13CH2][13CH2][13C](=O)[13C](=O)O
InChI key
KPGXRSRHYNQIFN-CVMUNTFWSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service