Skip to Content
Merck
All Photos(1)

Key Documents

86872

Sigma-Aldrich

Tetrabutylammonium fluoride trihydrate

≥97.0% (NT)

Synonym(s):

TBAF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]4NF · 3H2O
CAS Number:
Molecular Weight:
315.51
Beilstein:
3761900
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NT)

form

crystals

mp

62-63 °C (lit.)

functional group

amine

SMILES string

O.O.O.[F-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.FH.3H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;/h5-16H2,1-4H3;1H;3*1H2/q+1;;;;/p-1

InChI key

VEPTXBCIDSFGBF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium fluoride trihydrate is a mild base used in reactions like aldol-type condensation reactions, Michael-type reactions, ring-opening reactions. Its is also used as a promoter in cross-coupling reactions and cyclization of carbocycles and heterocycles.

Application

Reactant for:
Preparation of deprotecting agents in preparation of cellulose derivatives
Synthesis of lipophilic peptides for DNA transfections in vivo
Dehydrobromination reactions
Tetrabutylammonium fluoride trihydrate can be used as a base:
  • For the dehydrobromination of vinyl bromides to terminal acetylenes.
  • In the conversion of 1,1-dibromo-1-alkenes to terminal alkynes via Corey–Fuchs reaction.
  • In Hiyama cross-coupling reaction of aryl and heteroaryl chlorides with aryltrialkoxysilanes in the presence of a palladium catalyst.

It can be used to catalyze ethynylation of quinolines and isoquinolines using calcium carbide in aqueous N,N-dimethylacetamide.

Other Notes

Reagent for the cleavage of silyl ethers and other silyl protecting groups; Catalyst for various reactions with silicon compounds; Use as a base in organic synthesis; Instability of anhydrous TBAF

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Method for the Production of Terminal Alkynes from 1, 1-Dibromo-1-alkenes and its Application in the Total Synthesis of Natural Product Dihydroxerulin
Liu S, et al.
advanced synthesis and catalysis, 357(2-3), 553-560 (2015)
G. Majetich et al.
The Journal of Organic Chemistry, 51, 1745-1745 (1986)
An Efficient Method for the Production of Terminal Alkynes from 1, 1-Dibromo-1-alkenes and its Application in the Total Synthesis of Natural Product Dihydroxerulin
Liu S, et al.
Advanced Synthesis & Catalysis, 357(2-3), 553-560 (2015)
Ethynylation of Isoquinoline and Quinoline Derivatives with Calcium Carbide
Samzadeh-Kermani A
Synlett, 28(16), 2126-2130 (2017)
T.W. Greene et al.
Protective Groups in Organic Synthesis (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service