Skip to Content
Merck
All Photos(2)

Key Documents

861413

Sigma-Aldrich

γ-Cyclodextrin hydrate

Synonym(s):

γ-Schardinger dextrin, Cyclooctaamylose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H80O40 · xH2O
CAS Number:
Molecular Weight:
1297.12 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

optical activity

[α]20/D +176.1°, c = 1 in H2O (dried basis)

mp

267 °C (dec.) (lit.)

functional group

ether

SMILES string

O.OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]9[C@H](O)[C@@H](O)[C@H](O[C@@H]9CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C48H80O40.H2O/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58;/h9-72H,1-8H2;1H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-;/m1./s1

InChI key

SPKUKQHKKVTYOE-SMTXKKGASA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-cyclodextrin (γ-CD) is a water-soluble cyclic octamer of D-glucose units linked via α(1→4) glycosidic bonds. Its ability to form several inclusion complexes with organic as well as inorganic compounds makes it a potential candidate in solid and solution state determinations.

Application

γ-Cyclodextrin hydrate may be used to form inclusion complexes with steviol glycosides, 1-propanol, tempone and di-tert-butyl nitroxide followed by their characterization by various analytical techniques.
Modifier used in experiments evaluating viability, integrity, and inflammation in genital epithelia upon exposure to pharmaceutical excipients and microbicides

Reactant used to investigate host-guest interactions between α−, β−and γ-cyclodextrins and vanadocene dichloride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spin-label-induced selective carbon-13 nuclear relaxation: structures of the. gamma.-cyclodextrin-tempone and. alpha.-cyclodextrin-di-tert-butyl nitroxide inclusion complexes in solution
Eastman MP, et al.
Macromolecules, 22(10), 3888-3892 (1989)
Mani Upreti et al.
International journal of molecular sciences, 12(11), 7529-7553 (2011-12-17)
Steviol glycosidesrebaudioside (reb) A, C and D have low aqueous solubilities. To improve their aqueous solubilities, inclusion complex of steviol glycosides, reb A, C and D and gamma cyclodextrin were prepared by freeze drying method and further characterized by means
Structure of the gamma-cyclodextrin-1-propanol-17H2O inclusion complex
Ding J, et al.
Acta Crystallographica Section B, Structural Science, 47(5), 731-738 (1991)
Determination of the impurity profile of gamma-cyclodextrin by high-performance liquid chromatography
White G, et al.
Journal of Chromatography A, 625(2), 157-161 (1992)
beta-Cyclodextrin hydration: a calorimetric and gravimetric study
Bilal M, et al.
Thermochimica Acta, 249, 63-73 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service