Skip to Content
Merck
All Photos(1)

Key Documents

857327

Sigma-Aldrich

L-Cystine dimethyl ester dihydrochloride

≥95%

Synonym(s):

L-Cys(OMe)OMe·HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH(NH2)CH2SSCH2CH(NH2)COOCH3 · 2HCl
CAS Number:
Molecular Weight:
341.28
Beilstein:
4727524
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

optical activity

[α]22/D −36.0°, c = 4% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

182-183 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].Cl[H].COC(=O)[C@@H](N)CSSC[C@H](N)C(=O)OC

InChI

1S/C8H16N2O4S2.2ClH/c1-13-7(11)5(9)3-15-16-4-6(10)8(12)14-2;;/h5-6H,3-4,9-10H2,1-2H3;2*1H/t5-,6-;;/m0../s1

InChI key

QKWGUPFPCRKKMQ-USPAICOZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyu Zhang et al.
Biomaterials science, 7(5), 1962-1972 (2019-02-28)
Functional polymeric drug delivery systems have generated enormous interest due to their excellent features. This paper reports the development of a novel pH and redox dual-sensitive polymer for anticancer paclitaxel (PTX) delivery applications. The polymer was prepared by polycondensation of
Yu Huang et al.
Journal of materials chemistry. B, 9(7), 1888-1895 (2021-02-04)
It remains a major challenge to achieve efficient oral delivery and controlled intestinal release of ions using hydrogels. Herein, we report a novel, pH/redox-dual responsive, nanoemulsion-embedded composite hydrogel to address this issue. The hydrogel was first synthesized by crosslinking a
Manjusha Mathew et al.
Analytical biochemistry, 459, 31-38 (2014-05-20)
A highly sensitive electrochemical glucose sensor has been developed by the co-immobilization of glucose oxidase (GOx) and horseradish peroxidase (HRP) onto a gold electrode modified with biocompatible cyclic bisureas-gold nanoparticle conjugate (CBU-AuNP). A self-assembled monolayer of mercaptopropionic acid (MPA) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service