Skip to Content
Merck
All Photos(1)

Documents

808288

Sigma-Aldrich

Graphene dispersion

greener alternative

In water, inkjet printable

Synonym(s):

conductive ink, graphene ink, inkjetting ink

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

product name

Graphene ink in water, inkjet printable

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

~0.1 wt. % solids in water

sheet resistance

4k Ω/sq, 80 nm thickness

particle size

80-500 nm (exfoliated graphene flakes)

viscosity

1 cP (100s-1)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Graphene inks are highly conductive, low cost and super flexible. Click here for more information.

Application

Formulated for Inkjet printing:
  • Typical substates: plastic and glass.
  • Drying Condition: 100°C for 10 min.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory biomarkers in acute coronary syndromes: part I: introduction and cytokines.
Ehrin J Armstrong et al.
Circulation, 113(6), e72-e75 (2006-02-16)
A M Bofin et al.
Cytopathology : official journal of the British Society for Clinical Cytology, 14(6), 314-319 (2003-11-25)
The TOP2A gene is located on chromosome 17 close to the HER-2 gene. It encodes an enzyme involved in the regulation of cell proliferation. Using fluorescence in situ hybridization (FISH), we have examined fine needle aspiration smears from 42 cases
Line S Tarpgaard et al.
BMC cancer, 16, 91-91 (2016-02-13)
The overall purpose of this study is to provide proof of concept for introducing the anthracycline epirubicin as an effective, biomarker-guided treatment for metastatic colorectal cancer (mCRC) patients who are refractory to treatment with oxaliplatin-based chemotherapy and have TOP2A gene
H C van Zaanen et al.
The Journal of clinical investigation, 98(6), 1441-1448 (1996-09-15)
In vitro as well as in vivo observations have shown that IL6 plays a key role in the pathogenesis of multiple myeloma. Therefore we started a phase I/II dose escalating study with chimeric monoclonal anti-IL6 antibodies (cMab) in multiple myeloma
L M Biasucci et al.
Circulation, 99(16), 2079-2084 (1999-04-27)
A growing body of evidence suggests a role for inflammation in acute coronary syndromes. The aim of this study was to assess the role of proinflammatory cytokines, their time course, and their association with prognosis in unstable angina. We studied

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service