808083
Graphite
flakes, ≥98% carbon basis, -325 mesh particle size (50- 70%), natural
Synonym(s):
Graphite grade 3243
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Recommended Products
grade
natural
Quality Level
Assay
≥98% carbon basis
form
flakes
particle size
-325 mesh (44μ, 50-70%)
mp
3652-3697 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Flake graphite is a naturally occurring form of graphite that is typically found as discrete flakes ranging in size from 50-800 μm in diameter and 1-150 μm thick. This form of graphite has a high degree of crystallinity, which equates to near theoretical true density, high thermal and electric conductivity and low springback (excellent molding characteristics).
Application
- Graphene precursor
- Inorganic source of carbon
- Filler
- Thermal additive
- Re-carburizer
- Casting powders
- Drilling fluids
- Plastic additive
- Rubber additive
- Tint/pigment
- Lubricant
- Chemically resistant additive
- EMF absorber
- Milling and sieving
- General inert filler-additive
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Effects of different particles sizes of graphite on the engineering properties of graphites/polypropylene composites on injection molding aplication.
Iswandi, et al.
Key Engineering Materials, 109-114 (2011)
Preparation and properties of polypropylene nanocomposites reinforced with exfoliated graphene.
An, et al.
Fibers and Polymers, 13(4), 507-514 (2012)
Ethan B Secor et al.
The journal of physical chemistry letters, 6(4), 620-626 (2015-08-12)
Carbon and post-carbon nanomaterials present desirable electrical, optical, chemical, and mechanical attributes for printed electronics, offering low-cost, large-area functionality on flexible substrates. In this Perspective, recent developments in carbon nanomaterial inks are highlighted. Monodisperse semiconducting single-walled carbon nanotubes compatible with
Reverse-Micelle-Induced Exfoliation of Graphite into Graphene Nanosheets with Assistance of Supercritical CO2.
Xu, et al.
Chemistry of Materials, 27(9), 3262-3272 (2015)
Energy efficient thermal storage montmorillonite with phase change material containing exfoliated graphite nanoplatelets.
Jeong, et al.
Solar Energy Mat. and Solar Cells, 139, 65-70 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service