Skip to Content
Merck
All Photos(1)

Key Documents

791857

Sigma-Aldrich

Indium(III) phthalocyanine chloride

Dye content 95 %

Synonym(s):

Xerox xpp-ClInPc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16ClInN8
CAS Number:
Molecular Weight:
662.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, >95%
Dye content, 95%

mp

>300 °C (lit.)
>400 °C

λmax

697 nm

SMILES string

Cl[In]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.ClH.In/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;;/h1-16H;1H;/q-2;;+3/p-1

InChI key

AHXBXWOHQZBGFT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sensitizer for electrophotography

Legal Information

Product of Xerox®
Xerox® is a registered trademark of Xerox Corporation
Xerox is a registered trademark of Xerox Corporation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service