Skip to Content
Merck
All Photos(2)

Key Documents

791520

Sigma-Aldrich

Graphene oxide, ammonia functionalized

greener alternative

1 mg/mL, dispersion in H2O

Synonym(s):

N-doped GO, N-doped graphene oxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion in H2O

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1 mg/mL

greener alternative category

SMILES string

NC1=C2C3=C4C=C5C=C6C7=C8C9=C%10C%11=C%12C=C(O)C%13C%11C%14=C9C(N)(C7=O)c%15cc%16C%17=C(C(O)=O)C%18=CC%19=C%20C(=C1)C%21=C%22C%23=C%24C=C%25C%26=C%27C=C%28C%29=C%30C%31=C%32C%33=C%34C(C=CC(N)(C%28=C%35C(=O)C=C%36C%37OC%37%38C=C%39C=C%40C(=O)C%41(O)C=C%42C=C%43C=C(O)c%44cc(C(O)=O)c%45c%46c%44c%43c%47c%42c%48C%41=C%49C%40=C%50C(C%19=C%51C(=C%26C(=C%38C%51(N)C%39%50N)C%36%52OC%27%35%52)C%20%53OC%22%25%53)=C%18C%49%54OC%17%54c%48c%55c%16c%56c(c%57c%45ccc%13c%57c%14c%15%56)C%46%58OC%47%55%58)C%30%34N)=CC%59C(=O)C%60=CC%61=C(C(O)=O)c%62ccc(C(O)=O)c%63C=C%64C(=O)C%65%66OC%65%67C%68=C%64C(C%61C%69=C%60C(=C%33%59)C%70(N)C(=C%68%69)C%71=C%67C%72=C%73C%74=C%75C%76=C%77C%78(OC%78%79C=C%24C%80(N)C%81=C%23C%82(OC2%21%82)C%83=C3C%84=C(C(O)=C(O)C%85=CC%86=C(C(O)=O)C%87=CC%88=CC%89=C(C(O)=O)c%90ccc(O)c%12c%90C%10%91OC%89%91C8=C%88C6=C%87C5=C%86C4(N)C%84%85N)C%92%93OC%92C%94=C%95C(=O)C(O)=C%96C=C%97C%98C%99C%96=C%95C%100=C%101C%99=C%102C%103%104OC%103%105C%106=C%107C%108=C%109C(C%110=CC=C(O)C%111=CC%112=CC(=C%97C(O)=O)C(C%106=C%112C%108C%110%111)=C%98%104)=C(C=C%113C(=O)C%66=C%72C%114(OC%109%113%114)C%107=C%73C%105=C%75C%102%115OC%77%115C%101=C%79C%80(N)C%100=C%94C%81=C%83%93)C(O)=O)C(=C%29)C%31=C%76C(=C%71%74)C%32%70N)c%62%63

Looking for similar products? Visit Product Comparison Guide

General description

N-doped graphene may be prepared by chemical vapor deposition (CVD), direct synthesis of N-graphene and also by electrical annealing of graphene nanoribbons in NH3 in the presence of NH3.Another technique of preparing N-doped graphene by microbial reduction of graphene oxide was reported. N doping alters the electrical properties of graphene effectively by modifying its band structure. A report also shows that doped graphene can act as excellent support for Pt catalysts and show better performance in Li ion batteries.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". High concentrated graphene oxide sheets provide the prerequisite viscosity to bind the electrode materials together and enable 3D printing. Using water as a green solvent makes this aqueous ink system feasible for processing and drying safety and low cost. Click here for more information.

Application

Dielectric Materials for Field Effect Transistors (FET); Non-silicon-based materials for integrated circuits (IC).
Nitrogen doping (N-doping) can improve electro-conductivity of graphene sheet .

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of N-doped graphene by reduction of graphene oxide with mixed microbial system and its haemocompatibility
Fan M, et al.
Nanoscale, 6, 4882-4882 (2014)
Influence of N-doping on the structural and photoluminescence properties of graphene oxide films.
Khai TV, et al.
Carbon, 50, 3799- 3806 (2012)
Nitrogen-doped graphene sheets grown by chemical vapor deposition: synthesis and influence of nitrogen impurities on carrier transport.
Lu Y, et al.
ACS Nano, 7.8, 6522-6532 (2013)
Laser mediated programmable N doping and simultaneous reduction of graphene oxide.
Guo L,et al.
Advanced Optical Materials, 2, 120-125 (2014)
Chemical vapor deposition of N-doped graphene and carbon films: the role of precursors and gas phase.
Ito Y, et al.
ACS Nano, 8.4, 3337-3346 (2014)

Articles

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service