Skip to Content
Merck
All Photos(1)

Documents

790419

Sigma-Aldrich

Ethyl 2-ethoxy-2-iminoacetate

Synonym(s):

(1-Ethoxyformimidoyl)formic acid ethyl ester, 2-Ethoxy-2-iminoacetic acid ethyl ester, Ethyl 1-carbethoxyformimidate, Ethyl carboethoxyformidimate, Ethyl ethoxycarbonylformimidate, Ethyl ethoxyiminoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO3
CAS Number:
Molecular Weight:
145.16
UNSPSC Code:
12352100
NACRES:
NA.22

form

liquid

Quality Level

refractive index

n20/D 1.426

density

1.042 g/mL at 25 °C

storage temp.

−20°C

InChI

1S/C6H11NO3/c1-3-9-5(7)6(8)10-4-2/h7H,3-4H2,1-2H3

InChI key

DSHWMBCJDOGPTB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

162.3 °F

Flash Point(C)

72.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service