Skip to Content
Merck
All Photos(1)

Key Documents

778966

Sigma-Aldrich

(2-Hydroxypropyl)-β-cyclodextrin

produced by Wacker Chemie AG, Burghausen, Germany

Synonym(s):

(2-Hydroxypropyl)-beta-cyclodextrin, Cavasol® W7 HP, HP-β-CD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352201
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

Quality Level

form

powder

extent of labeling

molar substitution per 0.60

impurities

≤1.0% unsubstituted cyclodextrin
≤5.0% polypropylene glycol

ign. residue

≤2.5%

loss

≤7.0% loss on drying

SMILES string

CC(O)COCC1OC2OC3C(COCC(C)O)OC(OC4C(COCC(C)O)OC(OC5C(COCC(C)O)OC(OC6C(COCC(C)O)OC(OC7C(COCC(C)O)OC(OC8C(COCC(C)O)OC(OC1C(OCC(C)O)C2OCC(C)O)C(OCC(C)O)C8OCC(C)O)C(OCC(C)O)C7OCC(C)O)C(OCC(C)O)C6OCC(C)O)C(OCC(C)O)C5OCC(C)O)C(OCC(C)O)C4OCC(C)O)C(OCC(C)O)C3OCC(C)O

InChI

1S/C63H112O42/c1-22(64)8-85-15-29-50-36(71)43(78)57(92-29)100-51-30(16-86-9-23(2)65)94-59(45(80)38(51)73)102-53-32(18-88-11-25(4)67)96-61(47(82)40(53)75)104-55-34(20-90-13-27(6)69)98-63(49(84)42(55)77)105-56-35(21-91-14-28(7)70)97-62(48(83)41(56)76)103-54-33(19-89-12-26(5)68)95-60(46(81)39(54)74)101-52-31(17-87-10-24(3)66)93-58(99-50)44(79)37(52)72/h22-84H,8-21H2,1-7H3/t22?,23?,24?,25?,26?,27?,28?,29-,30-,31?,32?,33?,34?,35?,36?,37-,38?,39-,40+,41+,42+,43?,44+,45?,46+,47+,48+,49+,50+,51+,52-,53-,54-,55-,56-,57+,58-,59+,60-,61-,62-,63-/m1/s1

InChI key

ODLHGICHYURWBS-RYJYQAAZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(2-Hydroxypropyl)- β -cyclodextrin (HP-β-CD), is a macrocyclic oligosaccharide with a hydrophobic inner cavity and a hydrophilic outer surface, that allows it to incorporate hydrophobic drugs into the cavities, thus enhancing water solubility and bioavailability of poorly water-soluble drugs. It also possesses high surface activity and stabilizing performance.

Application

2-Hydroxypropyl)-β-cyclodextrin is used as a:
  • Macromolecular additive in the synthesis of single-phasetetragonal zirconium oxides.
  • Precursor in the synthesis of polyrotaxane.
  • Solubilizer and stabilizer in pharmaceutical formulations. HP-β-CD also shows potent anticancer activity.
The solubility of lipophilic drugs increases linearly with the concentration of hydroxypropyl-β-cyclodextrin (HBC) in aqueous solution because of the complex between HBC and the drug. This guest-host type complex is formed between the drug and the non-polar cavity in the HBC that results in enhanced solubility. Solutions may be lyophilized to produce freely soluble powders. Non-toxic in rabbits and mice.

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2-hydroxypropyl-$\beta$-cyclodextrin/pluronic-based polyrotaxanes via heterogeneous reaction as potential Niemann-Pick type C therapeutics
Yawo MA,et al.
Biomacromolecules, 14, 4189-4197 (2013)
?Back to the future?: a new look at hydroxypropyl beta-cyclodextrins.
Malanga M, et al.
Journal of Pharmaceutical Sciences, 105(9), 2921-2931 (2016)
2-Hydroxypropyl-?-cyclodextrin acts as a novel anticancer agent.
Yokoo M, et al.
PLoS ONE, 10(11), e0141946-e0141946 (2015)
Hydroxypropyl-$\beta$-cyclodextrin as a versatile additive for the formation of metastable tetragonal zirconia exhibiting high thermal stability
Lei B,et al.
CrystEngComm, 15, 2076-2083 (2013)
Hydroxypropyl-$\beta$-cyclodextrin-based solid dispersed granules: A prospective alternative to conventional solid dispersion
Jung Suk K,et al.
International Journal of Pharmaceutics, 628, 122286-122286 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service